Please login to your online account to display your discounted pricing

WS 12, Tocris

TRPM8 agonist; cooling agent

$163.07 - $678.85

Chemical Identifiers

Chemical Name or Material WS 12
Assay Percent Range >99%
Molecular Formula C18H27NO2
Formula Weight 289.41
Synonym (1R™, 2S™)-N-(4-Methoxyphenyl)-5-methyl-2-(1-methylethyl)cyclohexanecarboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

304010

 
r & d systems
RDS304010
10mg Each for $163.07

304050

 
r & d systems
RDS304050
50mg Each for $678.85
Description & Specifications

Specifications

Chemical Name or Material WS 12
CAS 68489-09-8
Recommended Storage Store at -20°C
Assay Percent Range >99%
Molecular Formula C18H27NO2
Formula Weight 289.41
Synonym (1R™, 2S™)-N-(4-Methoxyphenyl)-5-methyl-2-(1-methylethyl)cyclohexanecarboxamide
Quantity 10mg
Safety and Handling

Store at -20°C