missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Wright's Stain, Rapid Formula, for Staining and General Differentiation of Blood Corpuscles, Ricca Chemical
Supplier: Ricca Chemical Company RI93501
Specifications
7558-79-4 | |
BNIILDVGGAEEIG-UHFFFAOYSA-L | |
disodium;hydrogen phosphate | |
24203 | |
Characteristic alcohol | |
4 L |
HNa2O4P | |
OP(=O)([O-])[O-].[Na+].[Na+] | |
141.957 | |
CHEBI:34683 | |
Amber Glass Bottle |
Safety and Handling
Recommended Storage : Flammable