Please login to your online account to display your discounted pricing

Wright's Eosin Methylene Blue Solution, For Microscopy, EMD Millipore™


Manufacturer: emd millipore corporation  1013832500

 View more versions of this product

Catalog No. M1013832500

  • / Each
 

Description & Specifications

Chemical Identifiers

DOT Information DOT Class: 3 (6.1), Packaging Group: II
Chemical Name or Material Wright's Eosin Methylene Blue Solution
Physical Form Liquid
CAS 67-56-1
UN Number UN 1230
Health Hazard 1 H225: Highly flammable liquid and vapour.
H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled
H370: Causes damage to organs
Odor Smells like methanol
Packaging Plastic bottle
Appearance Blue liquid
CAS 107-21-1

Specifications

Chemical Name or Material Wright's Eosin Methylene Blue Solution
CAS 67-56-1
CAS 107-21-1
UN Number UN 1230
Accessories

H225: Highly flammable liquid and vapour.
H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled
H370: Causes damage to organs