Please login to your online account to display your discounted pricing

Wright-Giemsa Stain Mixture, Ricca Chemical

$126.42 - $230.52

Chemical Identifiers

Chemical Name or Material Wright-Giemsa Stain Mixture
CAS 67-56-1
CAS Min % 83.0
CAS Max % 86.0
CAS 56-81-5
CAS Min % 14.6
CAS Max % 15.19
CAS 68988-92-1
CAS Min % 0.32
CAS Max % 0.33
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

938032

 
ricca chemical company
RI938032
Glass amber 1L Each for $230.52

938016

  SDS
ricca chemical company
RI938016
Glass amber 500mL Each for $126.42
Description & Specifications

Specifications

Physical Form Liquid
Color Clear dark purple
Quantity 1L
Chemical Name or Material Wright-Giemsa Stain Mixture
CAS Max % 0.33
CAS 7558-79-4
CAS Min % 0.07
CAS 7778-77-0
CAS 67-56-1
CAS 56-81-5
CAS 51811-82-6
CAS Min % 0.23
CAS Max % 0.24
CAS Max % 0.07
CAS Min % 0.04
CAS Max % 0.04
CAS Min % 83.0
Packaging Glass amber
CAS Max % 86.0
CAS Min % 14.6
CAS Max % 15.19
CAS 68988-92-1
CAS Min % 0.32