Please login to your online account to display your discounted pricing

Wortmannin, Tocris


Potent, irreversible inhibitor of PI 3-kinase; Also inhibitor of PLK1

Quantity 1mg

Manufacturer: r & d systems  RDS12321

 View more versions of this product

Catalog No. 12321

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 428.44
Quantity 1mg
Chemical Name or Material Wortmannin
Synonym (1S,6bR,9aS,11R,11bR) 11-(Acetyloxy)-1,6b,7,8,9a,10,11,11b-octahydro-1-(methoxymethyl)-9a,11b-dimethyl-3H-furo[4,3,2-de]indeno[4,5,-h]-2-h]-2-benzopyran-3,6,9-trione
Molecular Formula C23H24O8
Assay Percent Range >98%
CAS 19545-26-7
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material Wortmannin
Assay Percent Range >98%
Molecular Formula C23H24O8
Formula Weight 428.44
Synonym (1S,6bR,9aS,11R,11bR) 11-(Acetyloxy)-1,6b,7,8,9a,10,11,11b-octahydro-1-(methoxymethyl)-9a,11b-dimethyl-3H-furo[4,3,2-de]indeno[4,5,-h]-2-h]-2-benzopyran-3,6,9-trione
Accessories

Desiccate at -20°C