missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Wortmannin, Penicillium funiculosum, ≥99%

Inhibitor of phosphatidylinositol 3-kinase

Manufacturer:  Alfa Aesar™ J63983MB

 View more versions of this product

Catalog No. AAJ63983MB



Specifications

Specifications

25mg
−30° to −10°C
Inhibitor of phosphatidylinositol 3-kinase
19545-26-7
428.44g/mol
Light sensitive
67676
White to Off-white
Powder
≥99%
C23H24O8
KY 12420
Signal Transduction Reagent
Powder
Documents