Worklon™ Reusable Polyester Cleanroom Coveralls
Comfortable coveralls of herringbone polyester.
Manufacturer:  Superior Uniform Group 1903M
 View more versions of this product
Catalog No. 01-352-50B
Description
Description
Resist mineral acids and alkalis
  • Fabric offers superior resistance to mineral acids and alkalis
  • Ideal for manufacturing, pharmaceutical and research facilities
  • Sturdy coil zipper runs entire length of front and fastens securely at neck
  • Stainless-steel snap fasteners on trouser bottoms, collar and cuffs for adjustment at ankle, neck and wrist
  • All seams double needled to prevent linting
Specifications
Specifications
Polyester
Medium
96.52 to 101.6 cm
Blue
38 to 40 in.