Please login to your online account to display your discounted pricing
Worklon™ Reusable Polyester Cleanroom Coveralls
Comfortable coveralls of herringbone polyester.
$68.78 - $107.25
Specifications
Material Polyester
View More Specs
Products
Catalog Number Mfr. No. Color Size Size (Metric) Chest Price Quantity    
01-352-50B
superior uniform group
1903M
Blue Medium 96.52 to 101.6cm Each for $68.78
01-352-50D
superior uniform group
1903XL
Blue X-Large 116.84 to 121.92cm Each for $68.78
01-352-52C
superior uniform group
1904M
White Medium 96.52 to 101.6cm Each for $99.28
01-352-52D
superior uniform group
1904L
White Large 106.68 to 111.76cm Each for $96.30
01-352-52E
superior uniform group
1904XL
White X-Large 116.84 to 121.92cm Each for $107.25
Description & Specifications
Specifications
Material Polyester
Resist mineral acids and alkalis
  • Fabric offers superior resistance to mineral acids and alkalis
  • Ideal for manufacturing, pharmaceutical and research facilities
  • Sturdy coil zipper runs entire length of front and fastens securely at neck
  • Stainless-steel snap fasteners on trouser bottoms, collar and cuffs for adjustment at ankle, neck and wrist
  • All seams double needled to prevent linting
ISO Class 5 (formerly Class 100) cleanroom compatible and shipped individually packaged in single polybags.