Worklon™ Reusable Polyester Cleanroom Coveralls
Comfortable coveralls of herringbone polyester.
Manufacturer: Superior Uniform Group 1904L
Description
Resist mineral acids and alkalis
- Fabric offers superior resistance to mineral acids and alkalis
- Ideal for manufacturing, pharmaceutical and research facilities
- Sturdy coil zipper runs entire length of front and fastens securely at neck
- Stainless-steel snap fasteners on trouser bottoms, collar and cuffs for adjustment at ankle, neck and wrist
- All seams double needled to prevent linting
Specifications
Polyester | |
Large | |
106.68 to 111.76 cm |
White | |
40 to 42 in. |