Worklon™ Reusable Polyester Cleanroom Coveralls
Comfortable coveralls of herringbone polyester.
Manufacturer: Superior Uniform Group 1904M
Description
Resist mineral acids and alkalis
- Fabric offers superior resistance to mineral acids and alkalis
- Ideal for manufacturing, pharmaceutical and research facilities
- Sturdy coil zipper runs entire length of front and fastens securely at neck
- Stainless-steel snap fasteners on trouser bottoms, collar and cuffs for adjustment at ankle, neck and wrist
- All seams double needled to prevent linting
Specifications
Polyester | |
Medium | |
96.52 to 101.6 cm |
White | |
38 to 40 in. |