Please login to your online account to display your discounted pricing

Worklon™ Footwear

Choose from three styles

$29.74 - $29.74

Specifications

Closure Type Elastic
Color White
Includes Rear gripper adjustment
Material Plastic, Nylon
Product Type Shoe Covers
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

01-352-37C

 
superior uniform group
1012L
Large Pair for $29.74
Description & Specifications

Specifications

Closure Type Elastic
Color White
Includes Rear gripper adjustment
Material Plastic, Nylon
Product Type Shoe Covers
For Use With (Application) Ideal for visitors and in-plant wear

Shoe Covers

  • Plastic-based taffeta with nylon fabric backing
  • Ideal for visitors and in-plant wear
  • Elasticized tops for snug fit
  • Nonskid soles
  • Gripper adjustment in rear
  • Color: white