Fisherbrand™ Wood Wall Cabinet, 18 in. Wide
Fisherbrand™ Wood Wall Cabinets bring quality, warmth and durability to your laboratory.
$2293.41 - $2427.24
Specifications
Cabinet Type Wall
Depth (English) 12 in.
Depth (Metric) 30.5 cm
Door Type Framed Glass Door
Height (English) 31.25 in.
View More Specs
Products 12
Catalog Number Mfr. No. Color Material Price Quantity  
Catalog Number Mfr. No. Color Material Price Quantity  
FB4564HM1
View Documents Promotion Details
N/A Natural Stain Maple Veneer
Each for $2,427.24
FB4564HM2
View Documents Promotion Details
N/A Honey Stain Maple Veneer
Each for $2,427.24
FB4564HM3
View Documents Promotion Details
N/A Amber Stain Maple Veneer
Each for $2,427.24
FB4564HM4
View Documents Promotion Details
N/A Auburn Stain Maple Veneer
Each for $2,427.24
FB4564HM5
View Documents Promotion Details
N/A Harvest Stain Maple Veneer
Each for $2,427.24
FB4564HM7
View Documents Promotion Details
N/A Mocha Stain Maple Veneer
Each for $2,427.24
FB4564HO1
View Documents Promotion Details
N/A Natural Stain Oak Veneer
Each for $2,293.41
FB4564HO2
View Documents Promotion Details
N/A Honey Stain Oak Veneer
Each for $2,293.41
FB4564HO3
View Documents Promotion Details
N/A Amber Stain Oak Veneer
Each for $2,293.41
FB4564HO4
View Documents Promotion Details
N/A Auburn Stain Oak Veneer
Each for $2,293.41
FB4564HO5
View Documents Promotion Details
N/A Harvest Stain Oak Veneer
Each for $2,293.41
FB4564HO7
View Documents Promotion Details
N/A Mocha Stain Oak Veneer
Each for $2,293.41
Description
Description
  • Made from hand selected premium rotary cut maple or rift cut oak veneers
  • Robotically applied environmentally friendly finishes are chemically resistant.
  • Wall Cabinets come with two adjustable shelves.
  • Framed Glass Doors will have tempered safety glass and have solid wood rails and stiles.
Compliance
SEFA 8
Specifications
Specifications
Wall
30.5 cm
31.25 in.
Right
2
18 in.
Cabinet
12 in.
Framed Glass Door
79.4 cm
1
2 years
45.7 cm
SDS
missing translation for 'documents'
missing translation for 'documents'