Learn More
Wood's metal, lump (approx. 100mm), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 388551000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver to Gray | |
70.0°C | |
Pb·Sn | |
100 g | |
Glass bottle |
Wood's metal | |
76093-98-6 | |
MFCD00198175 | |
647.28 | |
Rod |
Chemical Identifiers
76093-98-6 | |
Pb·Sn | |
MFCD00198175 |
Safety and Handling
GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Harmful if inhaled.
May damage the unborn child. Suspected of damaging fertility.
GHS P Statement
Obtain special instructions before use.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Avoid contact during pregnancy/while nursing.
GHS Signal Word: Warning
RUO – Research Use Only