Wing Top Accessory
Creates a fan-shaped flame for glass bending.
Manufacturer:  Fisher Science Education H6650
 View more versions of this product
Catalog No. S49125A
Specifications Description & Specifications
Specifications
0.51 in.
To create a fan shaped flame for glass bending
Brass
1.3cm
For burner tube of a bunsen burner (s49106)
Flame Spreader