missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

WIN 64338 hydrochloride, Tocris

Bradykinin B2 antagonist

$237.92 - $969.13

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C45H68ClN4OP·HCl
Formula Weight 783.95
Synonym (S)-4-[2-[Bis(cyclohexylamino)methyleneamino]-3-(2-naphthalenyl)-1-oxopropylamino]benzyl tributyl phosphonium chloride hydrochloride
Chemical Name or Material WIN 64338 hydrochloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
105710 R & D Systems
RDS105710
10mg Each for $237.92
105750 R & D Systems
RDS105750
50mg Each for $969.13
Specifications Description & Specifications

Chemical Identifiers

>98%
783.95
WIN 64338 hydrochloride
C45H68ClN4OP·HCl
(S)-4-[2-[Bis(cyclohexylamino)methyleneamino]-3-(2-naphthalenyl)-1-oxopropylamino]benzyl tributyl phosphonium chloride hydrochloride

Specifications

163727-74-0
C45H68ClN4OP·HCl
10mg
WIN 64338 hydrochloride
>98%
783.95
(S)-4-[2-[Bis(cyclohexylamino)methyleneamino]-3-(2-naphthalenyl)-1-oxopropylamino]benzyl tributyl phosphonium chloride hydrochloride

Safety and Handling

Recommended Storage : Desiccate at +4°C