Please login to your online account to display your discounted pricing
Bel-Art SP Scienceware™ WIlmad™ OmniFit™ Valve NMR System
Designed for easy injection of chemicals through a gas-tight syringe for air sensitive samples without using a glove box. Bel-Art SP Scienceware™ WIlmad™ OmniFit™ Valve NMR System is comprised of a 5mm end cap, 2-way valve, septum with cap, barrel coupler and a vacuum connector.
$42.97 - $454.31
Products
Catalog Number Mfr. No. Description Price Quantity    
16-800-137
bel-art sp scienceware™
660000037
Omni-Fit Septum Each for $42.97
16-800-138
bel-art sp scienceware™
660000038
Omni-Fit valve Each for $454.31
16-800-334
bel-art sp scienceware™
663060571
300mghz Omni-Fit NMR sample tube Each for $88.34
Description & Specifications
  • The Omni-Fit Tube consists of a 507-PP tube topped by a sturdy 2 in. section of medium-walled tubing which supports the valve system
  • Omni-Fit Sample tubes (507-OF-7) sold separately
  • Omni-Fit valve (OFV) is a complete accessory set
  • Additional septum (OF-30) are available in packages of 6
Due to the nature of glass, Extreme Caution should be exercised when using at elevated or reduced pressures as performance of tubes under pressure or vacuum cannot be guaranteed. Adequate safety shielding should always be used when working in these conditions.