missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Wilmad™ 4.0mm Solid State NMR Rotor Cap Remover

Eliminate possible damage of the cap and rotor with this zero turn cap remover. Bel-Art SP Scienceware™ Wilmad™ 4.0mm Solid State NMR Rotor Cap Remover simplifies removal of end caps and base plugs in solid state NMR.

Supplier:  Bel-Art™ 660000041

Catalog No. 16-800-141



Description

Description

  • For use with 4mm solid state caps
Specifications

Specifications

4 mm Rotor
Rotor Cap Remover
RS-301-4180
4.0mm Solid State NMR Rotor Cap Remover
Stainless Steel
SDS
Documents

Documents

Promotions

Promotions