missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

WIKI4, Tocris™

Tankyrase inhibitor; inhibits Wnt signaling

$250.58 - $1,042.86

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C29H23N5O3S
Formula Weight 521.59
Synonym 2-[3-[[4-(4-Methoxyphenyl)-5-(4-pyridinyl)-4H-1,2,4-triazol-3-yl]thio]propyl]-1H-benz[de]isoquinoline-1,3(2H)-dione
Chemical Name or Material WIKI4
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
485510 R & D Systems
RDS485510
10mg Each for $250.58
485550 R & D Systems
RDS485550
50mg Each for $1,042.86
Specifications Description & Specifications

Chemical Identifiers

>98%
521.59
WIKI4
C29H23N5O3S
2-[3-[[4-(4-Methoxyphenyl)-5-(4-pyridinyl)-4H-1,2,4-triazol-3-yl]thio]propyl]-1H-benz[de]isoquinoline-1,3(2H)-dione

Specifications

838818-26-1
C29H23N5O3S
10mg
WIKI4
>98%
521.59
2-[3-[[4-(4-Methoxyphenyl)-5-(4-pyridinyl)-4H-1,2,4-triazol-3-yl]thio]propyl]-1H-benz[de]isoquinoline-1,3(2H)-dione

Safety and Handling

Recommended Storage : Store at -20°C