Alfa Aesar™ Wig-L-Bug™ Accessories
For use with Digital Wig-L-Bug Mixing/Grinding Mill
Manufacturer:  Alfa Aesar™ 44530KM
 View more versions of this product
Catalog No. AA44530KM
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Polystyrene vial with polyethylene slip-on cap; 1 ft. long x 0.5 in. dia.
  • Polystyrene vial with polyethylene slip-on cap; 2 ft. long x 0.5 in. dia.
  • Adapter for 2 in. long, 0.5 in. diameter vials
Specifications
Specifications
Accessory for Digital Wig-L-Bug mixing/grinding mill
2 ft.
25/Pk.
Polystyrene Vial
Documents