Why Is the Water Green? Lab Investigation
Investigate how common pollutants affect pond microlife.
Manufacturer:  Fisher Scientific AZ201153
 View more versions of this product
Catalog No. S23194
Requires : Compound microscope (not included)
Description
Description
  • Learn how pollutants, detergents, and fertilizers impact the growth of microalgae in ponds and contribute to environmental problems
  • Use either included dried culture mix or actual pond water samples to investigate the effect of algae by phosphates and nitrates
  • Learn the concentrations at which these common pollutants cause algal blooms
  • Determine the effects of other common pollutants
Specifications
Specifications
Dried culture mix, test vials, microscope slides and cover slips, pipets, nitrate and phosphate pollutants, comprehensive teacher and student guides
40-Student class kit
Documents