Fisherbrand™ White Ribbed Polypropylene Caps with Pulp/Vinyl Liner
Commonly used for food, beverage, medical and chemical applications packed at less than 120° F. Fisherbrand™ White Ribbed Polypropylene Caps with Pulp/Vinyl Liner are known for good impact strength and resistant to mild acids, alkalis, solvents, alcohols, oils and aqueous products.
$121.33 - $558.93
Specifications
Liner Material Pulp/Vinyl
Closure Type Screw
Material Polypropylene
Color White
Closure Color White
View More Specs
Alert:
Not recommended for active hydrocarbons or bleaches.
Products ${productFamilyLength}
Catalog Number Mfr. No. Neck Finish Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Neck Finish Packaging Quantity Price Quantity  
02-911-844A Fisherbrand™
ALP263442
53-400 1300/Cs. Case of 1300 Each for $377.50
02-911-853A Fisherbrand™
ALP263443
70-400 760/Cs. Case of 760 Each for $347.77
02-912-088A Fisherbrand™
ALP263421L
70-400 Packed in 12 bags of 12 144 Each/Cs. Case of 144 Each for $121.33
02-912-291A Fisherbrand™
ALP263419L
48-400 Packed in 48 bags of 12 576 Each/Cs. Case of 576 Each for $558.93
02-912-292A Fisherbrand™
ALP263420L
53-400 Packed in 24 bags of 12 288 Each/Cs. Case of 288 Each for $304.30
02-911-804A Fisherbrand™
ALP263417L
20-400 576/Cs. Case of 576 Each for $139.80
02-912-286A Fisherbrand™
ALP263418L
28-400 576/Cs. Case of 576 Each for $148.23
Specifications
Description
Polypropylene caps are:
  • Known for good impact strength, durability, cost effectiveness, thermal resistance, and pliability
  • Autoclavable
Pulp/Vinyl liners are:
  • Made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
  • Commonly used for food, beverage, medical and chemical applications packed at less than 120° F., including mild acids, alkalis, solvents, alcohols, oils and aqueous products
Description & Specifications
Specifications
Pulp/Vinyl
Polypropylene
White
Screw
White