Please login to your online account to display your discounted pricing
Fisherbrand™ White Ribbed Polypropylene Caps with Pulp/Vinyl Liner
Commonly used for food, beverage, medical and chemical applications packed at less than 120° F. Fisherbrand™ White Ribbed Polypropylene Caps with Pulp/Vinyl Liner are known for good impact strength and resistant to mild acids, alkalis, solvents, alcohols, oils and aqueous products.
Manufacturer: fisherbrand™  ALP263443
 View more versions of this product
Pulp/Vinyl liner
 Alert
Not recommended for active hydrocarbons or bleaches.
Catalog No. 02-911-853A
  • / Case of 760 Each
Description & Specifications
Specifications
Product Type Polyvinyl Lined Cap
Closure Color White
Closure Material Polypropylene
Closure Type Screw
Color White
Material Polypropylene
Liner Material Pulp/Vinyl
Neck Finish 70-400
Quantity 760/Cs.
Shape Round
Polypropylene caps are:
  • Known for good impact strength, durability, cost effectiveness, thermal resistance, and pliability
  • Autoclavable
Pulp/Vinyl liners are:
  • Made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
  • Commonly used for food, beverage, medical and chemical applications packed at less than 120° F., including mild acids, alkalis, solvents, alcohols, oils and aqueous products