Fisherbrand™ White Ribbed Polypropylene Caps with Pulp/Vinyl Liner
Commonly used for food, beverage, medical and chemical applications packed at less than 120° F. Fisherbrand™ White Ribbed Polypropylene Caps with Pulp/Vinyl Liner are known for good impact strength and resistant to mild acids, alkalis, solvents, alcohols, oils and aqueous products.
Manufacturer:  Fisherbrand™ ALP263420L
 View more versions of this product
Catalog No. 02-912-292A
Alert:
Not recommended for active hydrocarbons or bleaches.
Includes: Pulp/Vinyl liner
Specifications
Description
Cap, Ribbed; Fisherbrand; White; Polypropylene; With Pulp/Vinyl Liner; 53-400; 288EA/Cs.
Polypropylene caps are:
  • Known for good impact strength, durability, cost effectiveness, thermal resistance, and pliability
  • Autoclavable
Pulp/Vinyl liners are:
  • Made of a vinyl coating applied to high density polyethylene coated paper and then it is laminated to pulpboard
  • Commonly used for food, beverage, medical and chemical applications packed at less than 120° F., including mild acids, alkalis, solvents, alcohols, oils and aqueous products
Description & Specifications
Specifications
Polypropylene
Polypropylene
288 Each/Cs.
White
53-400
Screw
White
Packed in 24 bags of 12
Cap
Pulp/Vinyl
Round