Fisherbrand™ White Polypropylene PE Foam/PTFE Cap
Economical alternative to Thermoset and Phenolic PTFE lined caps. Fisherbrand™ White Polypropylene PE Foam/PTFE Cap have a PTFE facing, backed by F217 liners.
$213.79 - $213.79
Specifications
Closure Material Polypropylene
Closure Liner PTFE/Polyethylene
Closure Type Screw Cap
Color White
Material Polypropylene
View More Specs
Products 1
Catalog Number Mfr. No. Closure Size Quantity Price Quantity  
Catalog Number Mfr. No. Closure Size Quantity Price Quantity  
02-912-076
View Documents Promotion Details
Fisherbrand™
014503
53-400 mm 100/Pk.
Pack of 100 Each for $213.79
Case of 5 Pack for $1,068.95
Description
Description
Polypropylene (PP) caps with F217 & PTFE liners are an economical alternative to Thermoset & Phenolic PTFE lined caps. They have a PTFE facing, backed by F217 liners. The F217 liners consist of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE). PTFE/F217 liners are very resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference.
  • Features F217 liners which consists of 3 plys: a low density foam core tightly sandwiched between two layers of polyethylene (LDPE)
  • PTFE/F217 liners are resilient and have strong resistance to chemicals and moisture and help keep bottle or jar contents free of odor and taste interference
Specifications
Specifications
Polypropylene
Screw Cap
Polypropylene
Sterile
PTFE/Polyethylene
White
Round
PE Foam/PTFE