Whirling Hygrometers
$170.10 - $174.15
Includes: Carrying case and plastic humidity chart
Products ${productFamilyLength}
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
S68613 Fisher Science Education
736700
Each for $170.10
S68614 Fisher Science Education
736710
Each for $174.15
Specifications
Description
Obtain fast and accurate humidity readings anywhere.
•    Compare reading of wet- and dry-bulb red liquid thermometers to a humidity chart
•    Thermometers are recessed into a protective plastic mounting that contains a water reservoir to eliminate the need to constantly rewet the wick
•    Choice of °C (-50° to +50°C) or °F (20° to 120°F) ranges
Order Info
Dimensions: 1L × 6W × 8 1/2 in.H (2.5 × 15.2 × 21.6cm)
4 oz. (108g) total weight
Warranty
Five-year manufacturer's warranty