Please login to your online account to display your discounted pricing

While You Were Out Series


Never leave your substitute high and dry again.

Manufacturer: fisher science education  2776

 View more versions of this product

Catalog No. S68106

  • / Each
 

Description & Specifications

Specifications

Product Type While You Were Out-Biology
Category Biology
For Use With (Application) Concept crossword puzzles, formula word searches, challenging games within each discipline, answer sheets
Format Book
Grade 9 to 12