Please login to your online account to display your discounted pricing

WHI-P 154, Tocris

JAK3 kinase inhibitor; Also inhibits EGFR

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material WHI-P 154
Assay Percent Range >99%
Molecular Formula C16H14BrN3O3
Formula Weight 376.2
Synonym 2-Bromo-4-[(6, 7-dimethoxy-4-quinazolinyl)amino]phenol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

311510

 
r & d systems
311510
10mg Each for $201.36

311550

 
r & d systems
311550
50mg Each for $844.73
Description & Specifications

Specifications

Formula Weight 376.2
Quantity 10mg
Chemical Name or Material WHI-P 154
Assay Percent Range >99%
Molecular Formula C16H14BrN3O3
CAS 211555-04-3
Synonym 2-Bromo-4-[(6, 7-dimethoxy-4-quinazolinyl)amino]phenol
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature