missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Where We Are in Space and Time
Hands-on activities emphasize observing cycles and calculating sizes of/distances to stars, planets, and galaxies.
$43.82 - $43.82
Specifications
Subject | Science |
---|---|
For Use With (Application) | Observing and measuring astronomical time, measuring astronomical distances using triangles and measuring astronomical distances using apparent brightness |
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
S01386
|
Fisher Science Education
PS12 |
Teachers Guide |
Each for $43.82
|
|
Description
Twenty-one activities cover three units: Observing and Measuring Astronomical Time, Measuring Astronomical Distances Using Triangles and Measuring Astronomical Distances Using Apparent Brightness.
Activity Book
- Students can write answers or calculations in the book; some pages can be cut out
- Lists required materials; includes a glossary and a key to identifying objects on the photographic plates
- Spiral-bound; 176 pages
- Gives answers to the homework questions and problems
- Offers suggestions on approaching each activity
- Includes black and white master prints
- 48 pages
Specifications
Science | |
Observing and measuring astronomical time, measuring astronomical distances using triangles and measuring astronomical distances using apparent brightness |