Please login to your online account to display your discounted pricing

Wheaton™ White Polyproylene Caps for Narrow-Mouth Containers


For use with Wheaton leak-resistant narrow mouth bottles

Manufacturer: wheaton™  23951602

 View more versions of this product

Catalog No. 09-877-14B

  • / Case of 72 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Redi-pak™ leak resistant narrow mouth bottle
Neck Finish 38-430
Material Polypropylene
Certifications/Compliance ISO 9001
Color White
No. per Case 72 Cs.