Please login to your online account to display your discounted pricing

Wheaton™ White Polypropylene Screw Caps with bonded PTFE Faced Silicone Liner for E-Z Ex-Traction™ Vials


PTFE faced silicone liner ideal for storage and containment of volatile solvents, sensitive compounds or corrosive chemicals

Manufacturer: wheaton™  WW240836

 View more versions of this product

Catalog No. 02-927-113

  • / Case of 100 Each
 

Description & Specifications

Specifications

Neck Finish 24-400
Autoclavable Yes
For Use With (Equipment) 20mL Vial
Certifications/Compliance ISO 9001
No. per Pack 100/Cs.
Material Polypropylene
Color White
Closure Liner PTFE/Silicone
No. per Case 100 Cs.

  • Autoclavable