DWK Life Sciences Wheaton™ White Polypropylene Screw Caps with bonded PTFE Faced Silicone Liner for E-Z Ex-Traction™ Vials
PTFE faced silicone liner ideal for storage and containment of volatile solvents, sensitive compounds or corrosive chemicals
Supplier: DWK Life Sciences W240833
Description
- Autoclavable
Specifications
PTFE/Silicone | |
White | |
1000 Cs. | |
15-425 | |
Autoclavable |
8mL Vial | |
Polypropylene | |
1000/Cs. | |
ISO 9001 |