Please login to your online account to display your discounted pricing

Wheaton™ White Polypropylene Screw Caps with bonded PTFE Faced Silicone Liner for E-Z Ex-Traction™ Vials


PTFE faced silicone liner ideal for storage and containment of volatile solvents, sensitive compounds or corrosive chemicals

Manufacturer: wheaton™  WW240831

 View more versions of this product

Catalog No. 02-927-108

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Closure Liner PTFE/Silicone
Color White
Material Polypropylene
No. per Case 1000 Cs.
No. per Pack 1000/Cs.
For Use With (Equipment) 2 and 4mL Vial
Autoclavable Yes
Neck Finish 13-425

  • Autoclavable