Please login to your online account to display your discounted pricing

Wheaton™ White Polypropylene Open-Top Caps w/PTFE Faced Silicone Liners for E-Z Ex-Traction™ Vials

Suitable for storage and containment of volatile solvents, sensitive compounds or corrosive chemicals

$266.66 - $1,423.72

Specifications

Certifications/Compliance ISO 9001
Closure Liner PTFE/Silicone
Autoclavable Yes
Color White
Material Polypropylene
View More Specs
Products
Catalog Number Mfr. No. No. per Case Neck Finish Price Quantity    

02-927-115

 
wheaton™
WW240840
250 Cs. 13-425 Case of 250 Each for $364.43

02-927-116

 
wheaton™
WW240841
1000 Cs. 13-425 Case of 1000 Each for $1,312.34

02-927-117

 
wheaton™
WW240842
250 Cs. 15-425 Case of 250 Each for $427.23

02-927-118

 
wheaton™
WW240843
1000 Cs. 15-425 Case of 1000 Each for $1,423.72

02-927-119

 
wheaton™
WW240844
100 Cs. 22-400 Case of 100 Each for $311.66

02-927-120

 
wheaton™
WW240845
500 Cs. 22-400 Case of 500 Each for $1,358.70

02-927-121

 
wheaton™
WW240846
100 Cs. 24-400 Case of 100 Each for $266.66

02-927-122

 
wheaton™
WW240847
500 Cs. 24-400 Case of 500 Each for $1,403.04
Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Closure Liner PTFE/Silicone
Autoclavable Yes
Color White
Material Polypropylene
For Use With Glass bottle or vial

  • Autoclavable

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.