Please login to your online account to display your discounted pricing

Wheaton™ White Polypropylene Open-Top Caps w/PTFE Faced Silicone Liners for E-Z Ex-Traction™ Vials


Suitable for storage and containment of volatile solvents, sensitive compounds or corrosive chemicals

Manufacturer: wheaton™  WW240843

 View more versions of this product

Catalog No. 02-927-118

  • / Case of 1000 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Glass bottle or vial
Autoclavable Yes
Neck Finish 15-425
No. per Pack 1000/Cs.
Color White
Certifications/Compliance ISO 9001
Material Polypropylene
Closure Liner PTFE/Silicone
No. per Case 1000 Cs.