DWK Life Sciences Wheaton™ White Polypropylene Open-Top Caps w/PTFE Faced Silicone Liners for E-Z Ex-Traction™ Vials
Suitable for storage and containment of volatile solvents, sensitive compounds or corrosive chemicals
Supplier: DWK Life Sciences W240842
Description
Suitable for storage and containment of volatile solvents, sensitive compounds or corrosive chemicals
- Autoclavable
Specifications
PTFE/Silicone | |
White | |
250 Cs. | |
15-425 | |
Autoclavable |
Glass bottle or vial | |
Polypropylene | |
250/Cs. | |
ISO 9001 |