Please login to your online account to display your discounted pricing

Wheaton™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners


Excellent for environmental samples, pharmaceuticals and diagnostic reagents

Manufacturer: wheaton™  239237

 View more versions of this product

Catalog No. 02-984-34

  • / Case of 72 Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Closure Liner PTFE/Polyethylene
Color White
For Use With Glass bottle or vial
Material Polypropylene
Neck Finish 38-400
No. per Case 72 Cs.