Please login to your online account to display your discounted pricing

Wheaton™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners


Excellent for environmental samples, pharmaceuticals and diagnostic reagents

Manufacturer: wheaton™  239235

 View more versions of this product

Catalog No. 02-984-26

  • / Case of 144 Each
 

Description & Specifications

Specifications

Neck Finish 28-400
For Use With (Equipment) Glass bottle or vial
Color White
Material Polypropylene
Certifications/Compliance ISO 9001
No. per Case 144 Cs.
Closure Liner PTFE/Polyethylene