Please login to your online account to display your discounted pricing

Wheaton™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners


Excellent for environmental samples, pharmaceuticals and diagnostic reagents

Manufacturer: wheaton™  239229

 View more versions of this product

Catalog No. 02-984-14

  • / Case of 144 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Glass bottle or vial
Neck Finish 20-400
Color White
Material Polypropylene
No. per Case 144 Cs.
Certifications/Compliance ISO 9001
Closure Liner PTFE/Polyethylene