missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
DWK Life Sciences Wheaton™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners
Excellent for environmental samples, pharmaceuticals and diagnostic reagents
$37.07 - $634.59
Specifications
For Use With (Equipment) | Glass bottle or vial |
---|---|
Color | White |
Material | Polypropylene |
Certifications/Compliance | ISO 9001 |
Catalog Number | Mfr. No. | Neck Finish | Material | No. per Case | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Neck Finish | Material | No. per Case | Price | Quantity | |||
02-984-14
|
DWK Life Sciences
239229 |
20-400 | Polypropylene | 144 Cs. |
Case of 144 Each for $114.68
|
|
|||
02-984-26
|
DWK Life Sciences
239235 |
28-400 | Polypropylene | 144 Cs. |
Case of 144 Each for $249.62
|
|
|||
02-911-233
|
DWK Life Sciences
239243 |
63-400 | Polypropylene | 48 Cs. |
Case of 48 Each for $369.73
|
|
|||
02-984-46
|
DWK Life Sciences
239241 |
53-400 | Polypropylene | 72 Cs. |
Case of 72 Each for $384.65
|
|
|||
02-984-22
|
DWK Life Sciences
239233 |
24-400 | Polypropylene | 144 Cs. |
Case of 144 Each for $139.56
|
|
|||
02-984-37
|
DWK Life Sciences
239238 |
43-400 | Polypropylene | 72 Cs. |
Case of 72 Each for $37.07
|
|
|||
02-984-40
|
DWK Life Sciences
239239 |
45-400 | Polypropylene | 72 Cs. |
Case of 72 Each for $276.97
|
|
|||
02-984-30
|
DWK Life Sciences
239236 |
33-400 | Polypropylene | 144 Cs. |
Case of 144 Each for $300.91
|
|
|||
02-984-18
|
DWK Life Sciences
239231 |
22-400 | Polypropylene | 144 Cs. |
Case of 144 Each for $130.12
|
|
|||
02-984-49
|
DWK Life Sciences
239242 |
58-400 | Polypropylene | 72 Cs. |
Case of 72 Each for $422.62
|
|
|||
02-984-34
|
DWK Life Sciences
239237 |
38-400 | Polypropylene | 72 Cs. |
Case of 72 Each for $186.41
|
|
|||
02-984-61
|
DWK Life Sciences
239246 |
89-400 | Polypropylene | 48 Cs. |
Case of 48 Each for $634.59
|
|
|||
02-984-55
|
DWK Life Sciences
239244 |
70-400 | Polypropylene | 48 Cs. |
Case of 48 Each for $444.50
|
|
|||
Description
Excellent for environmental samples, pharmaceuticals and diagnostic reagents
- PTFE faced foamed polyethylene liner offers chemical resistance
- Polyethylene foam provides compressibility and sealing properties
- Good for high purity chemicals, strong acids, and solvents
- For usewith glass bottles or vials
- Polypropylene
- Screw cap
- White
Specifications
Glass bottle or vial | |
White | |
Polypropylene | |
ISO 9001 |