DWK Life Sciences Wheaton™ White Polypropylene Caps with PTFE-Faced Foamed Polyethylene Liners
Excellent for environmental samples, pharmaceuticals and diagnostic reagents
Supplier: DWK Life Sciences 239242
Description
Excellent for environmental samples, pharmaceuticals and diagnostic reagents
- PTFE faced foamed polyethylene liner offers chemical resistance
- Polyethylene foam provides compressibility and sealing properties
- Good for high purity chemicals, strong acids, and solvents
- For usewith glass bottles or vials
- Polypropylene
- Screw cap
- White
Specifications
PTFE/Polyethylene | |
White | |
72 Cs. | |
ISO 9001 |
Glass bottle or vial | |
Polypropylene | |
58-400 |