Please login to your online account to display your discounted pricing

Wheaton™ White Polypropylene Caps with Polyvinyl Liners


Manufacturer: wheaton™  239216

 View more versions of this product

Catalog No. 02-984-42

  • / Case of 72 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Round bottle
Neck Finish 48-400
Material Polypropylene
Certifications/Compliance ISO 9001
Color White
No. per Case 72 Cs.
Closure Liner Poly/Vinyl