Wheaton™ White Polypropylene Caps with Polyvinyl Liners
Manufacturer:  Wheaton™ 239211
 View more versions of this product
Catalog No. 02-984-25
Specifications
Description
Cap, Polyropylene; Wheaton; White; Polyvinyl liner; Cap Finish: 28-400; Case of 144; Pack of 72
  • White polypropylene screw cap provides clear surface for writing on cap
  • Poly-vinyl liner provides an excellent moisture barrier
Description & Specifications
Specifications
ISO 9001
White
144 Cs.
Round bottle
Poly/Vinyl
Polypropylene
Polypropylene Caps
28-400