Please login to your online account to display your discounted pricing

Wheaton™ White Polypropylene Caps with Polyvinyl Liners


Manufacturer: wheaton™  239209

 View more versions of this product

Catalog No. 02-984-21

  • / Case of 144 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Round bottle
Neck Finish 24-400
Material Polypropylene
Color White
Certifications/Compliance ISO 9001
Closure Liner Poly/Vinyl
No. per Case 144 Cs.