DWK Life Sciences Wheaton™ White Polypropylene Caps with Polyvinyl Liners
Good for mild acids, alkalis, solvents, alcohols, oils and aqueous products
Supplier: DWK Life Sciences 239207
Description
- White polypropylene screw cap provides clear surface for writing on cap
- Poly-vinyl liner provides an excellent moisture barrier
Specifications
Polypropylene Cap | |
Round bottle | |
Polypropylene | |
22-400 |
Polyvinyl | |
White | |
144 Cs. | |
ISO 9001 |