Please login to your online account to display your discounted pricing

Wheaton™ White Polypropylene Caps with Foamed Polyvinyl Liners


Foamed polyethylene liner provides chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products

Manufacturer: wheaton™  239289

 View more versions of this product

Catalog No. 02-984-47

  • / Case of 72 Each
 

Description & Specifications

Specifications

For Use With (Equipment) Glass bottle or vial
Neck Finish 53-400
Color White
Certifications/Compliance ISO 9001
Material Glass
No. per Case 72 Cs.
Closure Liner Foam Polyethylene