DWK Life Sciences Wheaton™ White Polypropylene Caps with Foamed Polyvinyl Liners
Foamed polyethylene liner provides chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products
Supplier: DWK Life Sciences 239285
Description
- Liner provides tight seal
- Liner Material: Foam, Polyethylene
- Polypropylene
- Screw Cap
- White
Use these caps with glass bottles or vials
Order Info
Each pack contains 72
Specifications
Polyethylene Foam | |
White | |
72 Cs. | |
ISO 9001 |
Glass bottle or vial | |
Polypropylene | |
38-400 |