Please login to your online account to display your discounted pricing

Wheaton™ White Polypropylene Caps with Foamed Polyvinyl Liners


Foamed polyethylene liner provides chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products

Manufacturer: wheaton™  239281

 View more versions of this product

Catalog No. 02-984-24

  • / Case of 144 Each
 

Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Closure Liner Foam Polyethylene
Color White
Material Polypropylene
No. per Case 144 Cs.
For Use With (Equipment) Glass bottle or vial
Neck Finish 24-400