Please login to your online account to display your discounted pricing

Wheaton™ White Polypropylene Caps with Foamed Polyvinyl Liners

Foamed polyethylene liner provides chemical resistance for acids, alkalis, solvents, alcohols, oils, household cosmetics and aqueous products

$52.09 - $91.64

Specifications

Certifications/Compliance ISO 9001
Closure Liner Foam Polyethylene
Color White
For Use With Glass bottle or vial
View More Specs
Products
Catalog Number Mfr. No. No. per Case Material Neck Finish Price Quantity    

02-984-24

 
wheaton™
239281
144 Cs. Polypropylene 24-400 Case of 144 Each for $74.03

02-984-28

 
wheaton™
239283
144 Cs. Polypropylene 28-400 Case of 144 Each for $58.21

02-984-35

 
wheaton™
239285
72 Cs. Polypropylene 38-400 Case of 72 Each for $52.09

02-984-44

 
wheaton™
239288
72 Cs. Polypropylene 48-400 Case of 72 Each for $73.30

02-984-47

 
wheaton™
239289
72 Cs. Glass 53-400 Case of 72 Each for $81.12

02-984-50

 
wheaton™
239290
72 Cs. Polypropylene 58-400 Case of 72 Each for $83.00

02-984-56

 
wheaton™
239292
48 Cs. Polypropylene 70-400 Case of 48 Each for $91.64
Description & Specifications

Specifications

Certifications/Compliance ISO 9001
Closure Liner Foam Polyethylene
Color White
For Use With Glass bottle or vial

  • Liner provides tight seal
  • Liner Material: Foam, Polyethylene
  • Polypropylene
  • Screw Cap
  • White

Use these caps with glass bottles or vials

Each pack contains 72