DWK Life Sciences Volumetric Modules for Acura™ Electro 926 XS and 936 Single Channel Pipets
Interchangeable modules to fit different volumetric assemblies
Supplier: DWK Life Sciences W870016
Description
- For reduced shaft length and single channel macrovolume pipets
- One control unit fits a choice of 27 volumetric assemblies available separately
Specifications
±1.2% | |
Acura™ Electro™ 926 XS Reduced Shaft Length Single-Channel Pipette | |
Volumetric Module | |
Ergonomic | |
0.05μL |
200μL Pipette tip | |
1 | |
0.5 to 10 μL | |
Acura™ Electro™ 926XS reduced shaft length single channel pipette | |
Reduced Length |