Please login to your online account to display your discounted pricing
Wheaton™ Volumetric Modules for Acura™ Electro 926 Regular Shaft Length Pipets
Permanently store performance data in instrument's memory
Manufacturer: wheaton™  WW870112
 View more versions of this product
Catalog No. 03-410-179
  • / Case
Description & Specifications
Specifications
Number of Channels 1
Number of Channels 1
Length (English) Regular Length
Increments 0.01μL
Tip Style Ultra 10μL
Volume (Metric) Range 0.1 to 2μL
Volume (Metric) 0.1 to 2μL
Compatible Tips Ultra 10μL pipette tip
Finger Style Ergonomic
For Use With (Equipment) Acura™ Electro™ 926XS regular shaft length single channel pipette
Accuracy ±2.5%
Compatibility Ultra 10μL pipette tip
Capacity Range 0.1 to 2μL